DrainRack™ Jr. Microscope Slide Trays, Simport Scientific
Supplier: Simport Scientific
M710-50B M710-50G M710-50W M710-50Y M710-50P
CA48648-564CS 57.22 CAD
CA48648-564 CA48648-566 CA48648-570 CA48648-572 CA89039-732
DrainRack™ Jr. Microscope Slide Trays, Simport Scientific
Slide Holders
These rugged trays/drain racks can hold up to 100 microscope slides in 50 individual numbered slots
These microscope slide trays are made of plastic.
All slides are stored upright for easier insertion and removal. Simply tilt them forward or backward with one finger to easily pick up the desired slide. Bar codes can be read without having to remove the slides from the tray. Not autoclavable.