VWR® Floating Microtube Racks
Supplier: VWR International
HS2132AV HS2134AV
82024-504CS 310.06 CAD
82024-504 82024-510
VWR® Floating Microtube Racks
Racks Tube Racks
Polypropylene racks keep microtubes submerged in a water bath.
  • Polypropyelen construction
  • Detachable
  • Ideal for controlling sample temperature
Legs are detachable to allow tubes to be easily removed by pushing the rack down onto any surface.
Detachable legs provide two options:
Attach legs so rack stands upright on bench top
detach legs so samples can be quickly popped out by pushing down onto any surface
Caution: Not recommended for autoclaving.