NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Supplier: Avantor
4020.X00V0 4050.X00V0 4100.X00V0 4200.X00V0 NJ-4606701-02 NJ-4606710-02 NJ-4606728-02 NJ-4606736-02
80076-428CS 848.26 CAD
80076-428 53548-017 53548-019 53548-023 53548-025 80076-426 76459-484 76459-486 76459-488 76459-490
NORM-JECT® Luer Lock Sterile Syringes, Air-Tite Products
Syringes Syringes, Medical
NORM-JECT® syringes feature medical-grade plastics, including a polypropylene barrel and a polyethylene plunger. Great value at reasonable costs.
  • Sterile
  • Certified DNA free
  • Latex free
  • Rubber free
  • Silicone free
  • Styrene free
  • DEHP free
These syringes are the choice for any situation needing an inert, non-reactive syringe. Because of their composition, these syringes are indicated for Nuclear Medicine, Amniocentisis, IVF, embryo-transfer, chromatography and many laboratory procedures. They are more chemically resistant than rubber tipped syringes. These inert syringes do not contain contaminates often associated with disposables.
The syringes can be used for sampling, mixing, and filtering. They are smooth drawing with a positive safety stop to prevent accidental spills. They feature accurate, easy-to-read increments. Their dose saver design minimizes waste, saving up to 4.5 ml of medication per 100 syringes used, as compared to non low deadspace Luer syringes (Actual savings may vary).
Certifications: Meets ISO Standards, CE registered, FDA registered.
Ordering information: Sterile and individually wrapped.