SP Bel-Art ProCulture Round Microcentrifuge Floating Bubble Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18875-0400 F18875-1000 F18875-1600
60986-340CS 387.54 CAD
60986-340 60986-342 60986-358
SP Bel-Art ProCulture Round Microcentrifuge Floating Bubble Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Made especially for use with beakers and waterbaths, these space-saving racks are ideal for small quantities of tubes and for use in cell culture laboratories.
  • Buoyant TPX® brand polymethylpentene with tiny bubbles throughout for additional buoyancy
  • Insert tubes from the top and easily remove them by pressing the rack down on the bench
  • Withstands boiling for 30 minutes at 100 °C (212 °F)
  • Steam autoclavable at 121 °C (250 °F)
  • Leg length: 19 mm (3/4"); thickness: 6.4 mm (1/4")
Perfect for incubating microtubes in a beaker of water.
Order Now
SPECIFICATIONS
Environmentally Preferable