Monoject™ SoftPack Syringes, Sterile, Covidien
Supplier: Cardinal Health
1181621100 1181621100 Z1180600555 Z8881520673 Z1182000777 Z1183500777
82002-299CS 698.71 CAD
82002-299 CA82002-268 CA82002-299 CA82002-246 CA82002-278 CA82002-304 CA82002-306 CA82002-310
Monoject™ SoftPack Syringes, Sterile, Covidien
Syringes Syringes, Medical
These latex-free syringes feature a one-step peel-away package that facilitates quick and easy opening for aseptic handling.
The 3mL syringes feature extended finger flanges and a longer plunger rod for greater control. The 3mL, 6mL, and 12mL syringes feature bold graduations; the 35mL and 60mL syringes feature bold dual graduations. The 35mL syringes also feature 1oz. graduations in 0.25oz. increments, and the 60mL syringes have 2oz. graduations in 0.25oz. increments.
Each individual package contains a color-coded gauge stripe and bar code for easy gauge identification. SyringesMore Product Information