Control Slides, Azer Scientific
Supplier: Azer Scientific Inc.
ESCS-104 ESCS-106 ESCS-107 ESCS-109 ESCS-111 ESCS-113 ESCS-114 ESCS-115 ESCS-116 ESCS-117 ESCS-118 ESCS-121 ESCS-122 ESCS-124 ESCS-125 ESCS-126 ESCS-127 ESCS-129 ESCS-130
95042-282PK 222.25 CAD
95042-282 95042-286 95042-288 95042-292 95042-296 95042-300 95042-302 95042-304 95042-306 95042-308 95042-310 95042-316 95042-318 95042-322 95042-324 95042-326 95042-328 95042-332 95042-334
Control Slides, Azer Scientific
Slides Microscopy Control Slides
Control slides are designed for use in monitoring stain performance in the Pathology lab.
  • Permanent positive charge for improved readability
  • Control slides with paraffin-embedded human tissue sections
  • Ten unstained Unifrost slides
  • One stained slide
All slides feature a permanent positive charge that securely anchors all types of sections and improves the quality of preparations and readability
Includes 10 unstained Unifrost microscope slides and one additional slide, which is stained with the proper procedure to demonstrate and validate the expected result.
Azer Scientific's control slides are carefully prepared with paraffin-embedded human tissue sections.
Packaging: 11 per pack