Nalgene® CryoBox™ Racks, Stainless Steel, Thermo Scientific
Supplier: Thermo Fisher Scientific
DS5035-0004 DS5035-0009 DS5037-0004 DS5037-0007 5036-0004
55710-107EA 563.13 CAD
55710-107 55710-129 55710-167 55710-169 55710-140
Nalgene® CryoBox™ Racks, Stainless Steel, Thermo Scientific
Racks Cryogenic Box Racks
Vertical racks for chest-type freezers.
Innovative retainer holds each box securely but separately. Allows fast retrieval of one box and immediate return of rack to freezer. Racks accommodate 5x5 vial (see 55709-946) or 9x9 vial (see 55709-968) NALGENE® CryoBox™ Boxes, and cardboard or stainless steel storage boxes with similar dimensions.