AzerMark™ Etching Slides, Azer Scientific
Supplier: Azer Scientific
AzerMark™
EMS300BW
89087-558CS 787.28 CAD
89087-558
AzerMark™ Etching Slides, Azer Scientific
Slides Microscope Slides
These slides are designed for use with automated slide etch labeling machines.
  • White glass with ground edges
  • Black and white labeling area
  • Dimensions: 75 x 25 x 1 mm
  • Adhesion available
The slides have white glass with ground edges, and the frosted coating retains most laboratory marking methods.
They are available with or without adhesion and include a black and white labeling area.
Ordering information: Slides are packaged 72 per box, 20 boxes per case (10 gross).