Barnstead/Thermolyne Cryogenic Freezer Racks for Mechanical Freezers, Thermo Scientific
Supplier: Thermo Fisher Scientific
HR509X7A HR509X5A
55709-403EA 682.24 CAD
55709-403 55709-425 55709-447 55709-469 55709-527
Barnstead/Thermolyne Cryogenic Freezer Racks for Mechanical Freezers, Thermo Scientific
Racks Cryogenic Box Racks
These storage systems are for use with 2.0mL or smaller ampules
Ideal for use with 81-ampule, 5.1cm (2") high cardboard and stainless steel boxes. Racks also hold 81- or 25-ampule NALGENE® CryoBox™ boxes (see 55709-946 series in Cryogenic Storage section). They are constructed of heavy duty stainless steel. Each rack unit is equipped with stainless steel retaining rod, which anchors boxes in the rack.