Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Supplier: Erie Scientific
7951PLUS-006 6951PLUS-006 8951PLUS-006
48312-906CS 2169.58 CAD
48312-906 48312-903 48312-905 48312-909
Colorfrost® Plus Microscope Slides, Erie Scientific (Thermo Scientific)
Slides Microscope Slides
A permanent positive charge applied to the slide surface allows slides to hold tissue sections without the need for expensive or time-consuming preparatory steps in the laboratory
This treatment virtually eliminates tissue loss during staining and electrostatically attracts frozen tissue sections and cytology preparations, binding them to the slide. Color-coded for quick and simple identification.
Order Now
SPECIFICATIONS
Environmentally Preferable