Epredia™ Superfrost™ Plus Control Slides
Supplier: Erie Scientific
ES-334PLUS-001 ES-3012-GLPLUS-001
48404-474PK 17.68 CAD
48404-474 48404-476 48404-478 48404-480
Epredia™ Superfrost™ Plus Control Slides
Slides Microscopy Control Slides
Ideal for staining procedures that require a control, these frosted slides feature spaces for both the patient sample and the control material
Slides may be used in histology, cytology, or microbiology applications. Slides are designed to ensure that tissue samples remain on the slide. A red or gold box identifies the control section. A red control box on the back surface identifies slides intended for use in automated instruments. Plus Gold control slides are recommended for hard-to-hold sections.