Clipped Corner Microscope Slides, Epredia™
Supplier: Epredia
4951-4-001 4941-4-006 4951-4-001
89080-574CS 1007.38 CAD
89080-574 56700-174 CA56700-174
Clipped Corner Microscope Slides, Epredia™
Slides Microscope Slides
Slides feature consistent 45° clipped corners for ease of handling and to aid with manual blood smears
Offering exceptional cleanliness and resistance to sticking, these slides are ideal for technicians who routinely prepare large numbers of slides. Superfrost® slides have an opaque coating that takes and retains most laboratory marking methods better than plain or conventional frosted slides. The Superfrost® coating is impervious to most common laboratory chemicals and reagents.