Cryo Boxes, PC
Supplier: NEST SCIENTIFIC
616001 616121 616204 616224 616305 616325 616421 616521 616931 616932
77508-362CS 511.18 CAD
77508-362 77508-366 77508-368 77508-370 77508-372 77508-374 77508-376 77508-378 77508-380 77508-382
Cryo Boxes, PC
Boxes Cryoboxes
Cryo boxes are manufactured in 100,000 grade cleaning room with USP 6 grade raw materials.
  • Temperature range: −196 to +70 ℃
  • Individual package
  • Clear polycarbonate cover for easy identification
  • Alphanumeric grid for easy sample identification
  • Each box includes drain holes and air vents to minimize condensation