Type-K Thermocouple Probes, Sper Scientific
Supplier: Sper Scientific
800061 800062 800064 800065 800066 800072 800073 800077 800060 800070
14003-116EA 215.91 CAD
14003-116 14003-118 14003-120 14003-122 14003-124 14003-130 14003-132 14003-136 14224-534 14003-126
Type-K Thermocouple Probes, Sper Scientific
Temperature Probes
Type-K thermocouple probes for use with Type-K thermometers in 14003-070 series.
Ambient probes provide fast response in air or gas. Choose the beaded wire ambient probe to sample hard-to-reach areas.
Ordering information: Type-J probes are available; contact your representative for information.