Four-Neck Reaction Vessel Lids, Chemglass
Supplier: Chemglass
CG-1941-01 CG-1941-02 CG-1941-03 CG-1941-04 CG-1941-05 CG-1941-06 CG-1941-07 CG-1941-08 CG-1941-09 CG-1942-01 CG-1942-02 CG-1942-03 CG-1942-05
47733-862EA 1136.45 CAD
47733-862 47733-864 47733-866 47733-868 47733-870 47733-872 47733-874 47733-876 47733-878 80071-584 80071-586 80071-588 80071-592
Four-Neck Reaction Vessel Lids, Chemglass
Lids Reaction Vessel Lids
These four neck reaction vessel lids have a flat ground surface allowing it to be greased or used with an o-ring.
Lids with thermometer side neck have a 10/30 outer joint which will permit a thermometer to be inserted vertically to the bottom of the vessel.
Ordering information: Four neck lids are available with joint sizes and configurations other than those listed.