Corning® Frosted Micro Slides, Corning
Supplier: Corning
2949-75X25 2948-75X25
48312-048CS 1549.05 CAD
48312-048 89091-004
Corning® Frosted Micro Slides, Corning
Slides Microscope Slides
For use in nonsterile cytology and pathology applications.
Made of water-white glass for maximum clarity. Precleaned and ready for use. Inspected for chips and rough edges before packaging. Slides are sandblasted and not etched, resulting in high legibility against most laboratory bench backgrounds.
Ordering information: Packaged 1/2 gross per box. Slides are packed vertically for easy lift out.