VWR® Premium Superfrost® Plus Microscope Slides
Supplier: VWR
VWR® Superfrost®
4951PLUS-601640 4951PLUS
CA48311-703CS 0 CAD
CA48311-703 48311-703
VWR® Premium Superfrost® Plus Microscope Slides
Slides Microscope Slides
The VWR® Superfrost® Plus micro slide features a special treatment that eliminates the need for additional adhesives or protein coatings, and improves tissue adherence in microwave stainings.
  • Ideal for frozen tissue sections and cytology preparation applications
  • Opaque coating reliably retains laboratory markings
  • Positively charged
Slides are specially treated for frozen tissue sections and cytology preparation applications, eliminating the need for special adhesives and protein coatings. Cytology preparations placed on these slides resist cell loss during enzyme digestion, DNA denaturation, and RNA hybridization. Slides also improve tissue adherence during microwave staining. Slides have an opaque coating that retains most laboratory marking methods.