Airfree® Reaction Flasks, Kjeldahl Shaped, Chemglass
Supplier: Chemglass
AF-0520-03 AF-0520-01 AF-0520-02 AF-0520-04 AF-0520-05 AF-0520-06 AF-0520-07 AF-0520-08
80064-590EA 278.56 CAD
80064-590 80064-586 80064-588 80064-592 80064-594 80064-596 80064-598 80064-600
Airfree® Reaction Flasks, Kjeldahl Shaped, Chemglass
Reaction Vessels Reaction Vessels, Flask Shape
Kjeldahl shape facilitates heating and stirring while permitting easy transfer of liquids or solids
Lower portion of flask fits standard heating mantles. Comes with a 2mm glass stopcock, constant pressure retaining device, and an all fluoropolymer resin AIRFREE® valve.