Pierce™ Immunoprecipitation, Protein A/G Coated Plate IP Kit, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce
45350 45350
CAPI45350EA 1011.83 CAD
CAPI45350 PI45350
Pierce™ Immunoprecipitation, Protein A/G Coated Plate IP Kit, Thermo Scientific
Protein Purification Protein Purification Kits
Pierce™ Protein A/G Coated Plate IP Kit uses coated Protein A/G microplates to perform immunoprecipitation assays in 96-well plates without resins, beads, centrifugation or magnets.
  • Ready-to-use, high quality coated plates provide high capacity and consistency
  • Plate format best suited for simultaneously processing multiple samples and their control conditions
  • Faster, easier and more thorough washing than with traditional tube/resin IP methods
  • Uses familiar and convenient ELISA tools (multichannel pipettors and plate washing); no tedious separation of supernatant from pelleted resin beads, and no tubes to open and close and centrifuge
Pierce Coated Plate IP Kits enable rapid immunoprecipitation of multiple samples without the usual tedium of pipetting, centrifuging and separating beaded affinity resin in individual microcentrifuge tubes.
Delivery information: Kit includes Protein A/G Coated 96-Well Strip Plates (2 Plates), Uncoated 96-Well Strip Plate, White (2 Plates), PBS Pack (makes 500 mL), Surfact-Amps™ X-100 Detergent Solution (6x10 mL), Elution Buffer, pH 2.8 (50 mL), Neutralization Buffer (12 mL) and Sealing Tape for 96-Well Plates (18 Sheets).