CellPro™ Cryovial, Externally Threaded Caps, PP, Sterile, Self Standing with NoLeak™ Technology
Supplier: Alkali Scientific
VCE-24 VCE-25 VCE-38
104028-330EA 530.77 CAD
104028-330 104027-006 104028-080
CellPro™ Cryovial, Externally Threaded Caps, PP, Sterile, Self Standing with NoLeak™ Technology
Vials
Ensure secure storage of your samples with CellPro™ cryovials featuring externally threaded caps made of PP, designed with sterile, self-standing, and NoLeak™ technology.
  • Sterilized by E-Beam
  • Ultra low temperature
  • Non-pyrogenic, DNase/RNase and human DNA free
  • Externally threaded caps
  • Round bottom, self-standing
CellPro™ Cryovials are designed for safe storage of samples at temperatures as low as −196 °C (gas phase of liquid nitrogen only). Cryogenic vials with both internally and externally threaded caps have a special screw cap designed with silicone O-rings to avoid the risk of sample contamination. The silicone O-rings allow for expansion and contraction in liquid nitrogen environments associated with long term, −190 °C storage.