PureClean™ Tyvek® 1421B Autoclavable Muff Covers
Supplier: KEYSTONE ADJUSTABLE CAP CO., INC.
10TK03 10TK04 10TK05 10TK06 10TK07 10TK08 10TK09 10TK12 10TK16 10TK18
77531-078CS 2867.7 CAD
77531-078 77531-062 77531-068 77531-064 77531-074 77531-070 77531-076 77531-080 77531-066 77531-072
PureClean™ Tyvek® 1421B Autoclavable Muff Covers
Covers
PureClean™ Autoclavable Muff Covers are made from 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilization applications. Keystone's PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Uncoated Tyvek® for maximum breathability during sterilization
  • Eliminate the risk of residue deposits on equipment post sterilization
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces labor costs and simplifies the conformance to wrapping SOP's
  • Autoclavable at 121 to 125 °C
  • Tear- and puncture-resistant, hydrophobic, lint-free
  • Conveniently packaged, 100 per sealed poly bag
PureClean™ Tyvek® Muffs provide a convenient means to cover flasks, beakers, containers, media bottles, valves, pipes, and hoses. These covers are breathable and can be used to prevent particulates and microbial contamination from entering openings of any size. Since they are autoclavable, they can be used during sterilization to reduce the time-consuming wrapping of open containers. Each closure incorporates latex-free rubber elastic to create a positive seal around round shaped openings.