Silicone Sealing Mats
Supplier: NEST SCIENTIFIC
506004 506005 506006 506016 506001 506002 506102 506162
77508-170CS 329.19 CAD
77508-170 77508-172 77508-174 77508-176 77508-164 77508-166 77508-190 77508-192
Silicone Sealing Mats
Seals Microplate Seals
The product is easy to open and close, and can effectively seal the deep well plate, which is convenient for operation and saves time.
Good sealing performance can prevent the volatilization of a variety of solvents. Chemical corrosion resistance and is used for low-temperature preservation. The pre-opening design allows the pipette tip to enter the hole without damage to the silicone sealing mat cover.