PIG® Barrel Top Absorbent Mat, New Pig
Supplier: New Pig
MAT208 MAT244 MAT237 MAT255
56620-036CS 242.23 CAD
56620-036 56620-678 89073-834 89073-836
PIG® Barrel Top Absorbent Mat, New Pig
Spill Control Materials
Soak up pump drips without cluttering up your work space with dirty rags
  • Absorbs and retains oils, coolants, solvents, and water
  • Dark gray color hides grime, so mat stays on the job longer
  • Fits 55 gallon steel drum tops; pre-cut with holes for 3/4" and 2" bungs to provide a perfect fit
  • Dispenser box makes it easy to carry and store mats while protecting them from dirt and moisture
  • Heavy weight construction is highly durable and absorbent to handle pump drips and dispensing spills
Eight layers of 100% polypropylene are thermally bonded to make PIG MatMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable