VWR VistaVision™ Cover Glasses, No. 2
Supplier: VWR International
VWR® VistaVision™
200252 122222 200222 200182 122525 121818
16004-316CS 352.22 CAD
16004-316 16004-304 16004-334 16004-324 16004-320 16004-342
VWR VistaVision™ Cover Glasses, No. 2
Cover Glasses
Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness, and are free of bubbles and scratches.
  • Pure white borosilicate glass fabrication
  • Excellent chemical resistance and flatness
Under ultraviolet light, they emit no disturbing self-fluorescence. Suitable for use in coverslippers. Manufactured in accordance with DIN ISO 8255-1.
Refractive index: 1.525 ±0.0015 ne
Ordering information: Square and circular cover glasses are supplied 1 oz. per two-part box.