VWR VistaVision™ Cover Glasses, No. 1
Supplier: VWR International
112222 112460 112450VWR 200181 112440 111818 200251 112250 112240 112525 112260 112230 200221 112430 112460A
16004-094CS 544.68 CAD
16004-094 16004-096 16004-098 16004-300 16004-306 16004-308 16004-310 16004-314 16004-318 16004-328 16004-330 16004-332 16004-338 16004-346 89218-832
VWR VistaVision™ Cover Glasses, No. 1
Cover Glasses
Microscope cover glasses are made of pure white borosilicate glass with excellent chemical resistance and flatness, and are free of bubbles and scratches.
  • Pure white borosilicate glass fabrication
  • Excellent chemical resistance and flatness
Under ultraviolet light, they emit no disturbing self-fluorescence. Suitable for use in coverslippers. Manufactured in accordance with DIN ISO 8255-1.
Refractive index: 1.525 ±0.0015 ne
Ordering information: Circular and square cover glasses are supplied 100 per two-part box; rectangular cover glasses are supplied 100 per hinged boxes.