VWR® SuperSlips™ Micro Cover Glasses, Rectangular, No. 1
Supplier: VWR International
VWR® SuperSlips™
VWR24X60-1WYOG VWR24X40-1WYOG 24X40-1W-YO-601640 VWR24X50-1WYOG
CA48395-090-1CS 166.71 CAD
CA48395-090-1 48404-457 48404-455 48404-453
VWR® SuperSlips™ Micro Cover Glasses, Rectangular, No. 1
Cover Glasses
These cover glasses are corrosion-resistant and uniformly thin and flat.
  • Uniformly thin and flat
  • Suitable for humid environments
  • Resistant to corrosion
The processing techniques result in superior cleanliness. Superslip coverslips combine specialized production methods with moisture-resistant packaging to prevent sticking. Suitable for applications in humid environments. Thickness: 0.13 to 0.17 mm. Supplied in 1 oz. increments.