Sleeve-Style Rubber Stoppers, Wheaton
Supplier: DWK Life Sciences (KIMBLE)
224100-060 224100-161 224100-020 224100-160 224100-320
80093-832CS 1467.38 CAD
80093-832 80093-834 89047-004 89047-006 16170-462
Sleeve-Style Rubber Stoppers, Wheaton
Stoppers Rubber Stoppers
Plug stoppers with fold-over skirts are for use on serum vials and other crimp-finish containers that do not require an aluminum seal or overcap
Stoppers may also be used as injection sites for multiple dose entry. General-purpose natural red rubber is excellent for resealing and multi-puncture applications. Diameter: 100mm.