VWR® One-Hole Kjeldahl Rubber Stoppers, Black
Supplier: VWR International
59587-400 59587-501
59587-400CS 703.22 CAD
59587-400 59587-501
VWR® One-Hole Kjeldahl Rubber Stoppers, Black
Stoppers Rubber Stoppers
These black rubber, one-hole stoppers are specifically designed to fit 500 and 800 mL Kjeldahl nitrogen determination flasks.
  • Excellent resiliency
  • Very pliable stoppers
  • Stoppers come with an 8mm-diameter hole
Stoppers have low free-sulfur content (1/2%), lower density than neoprene, and excellent resiliency and pliability. Both sizes come with an 8mm diameter hole.