PYREX® PTFE [ST] Keyhole Stoppers, Corning
Supplier: Corning
7630-13 7630-16 7630-19 7630-22
59501-032CS 165.58 CAD
59501-032 59501-033 59501-034 59501-035
PYREX® PTFE [ST] Keyhole Stoppers, Corning
Stoppers PE/LDPE/HDPE Stoppers
PTFE. Ideal for flasks that are grounded to [ST] stopper dimensions.
  • The color-coded keys make sorting and selecting the correct size faster and easier
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification I-RECs, ISO 14001 - Monterrey