PYREX® Hollow [ST] Reagent Bottle Stoppers, Corning
Supplier: Corning
7575-14 7575-19 7575-29 7575-24
89091-604CS 201.24 CAD
89091-604 89091-606 59435-660 89091-608
PYREX® Hollow [ST] Reagent Bottle Stoppers, Corning
Stoppers Glass Stoppers
Combination glass stoppers with closed bottoms may be used as [ST] joint stoppers and as reagent bottle stoppers
They are lightweight, but strong enough to keep chipping and breakage to a minimum. Barrel-shaped head is easy to clean and rotate, side grooves prevent slippage, and flat top allows stopper to stand on head.