VWR® Corks
Supplier: VWR International
CA23420-023 CA23420-038 CA23420-060 CA23420-071 CA23420-082 CA23420-107 CA23420-130 CA23420-140 CA23420-151 CA23420-195 CA23420-210 CA23420-231 CA23420-300 CA23420-388 CA23420-424 CA23420-468 CA23420-570 CA23420-050 CA23420-220 CA23420-242 CA23420-344 CA23420-366 CA23420-399 CA23420-413 CA23420-435 CA23420-457 CA23420-479 CA23420-480 CA23420-490 CA23420-515 CA23420-537 CA23420-548 CA23420-559 CA23420-560 23420-118 23420-162 23420-184 23420-300
CA23420-023PK 59.68 CAD
CA23420-023 CA23420-038 CA23420-060 CA23420-071 CA23420-082 CA23420-107 CA23420-130 CA23420-140 CA23420-151 CA23420-195 CA23420-210 CA23420-231 CA23420-300 CA23420-388 CA23420-424 CA23420-468 CA23420-570 CA23420-050 CA23420-220 CA23420-242 CA23420-344 CA23420-366 CA23420-399 CA23420-413 CA23420-435 CA23420-457 CA23420-479 CA23420-480 CA23420-490 CA23420-515 CA23420-537 CA23420-548 CA23420-559 CA23420-560 23420-118 23420-162 23420-184 23420-300
VWR® Corks
Stoppers Cork Stoppers
XXXX Corks function as a closure for bottles, vials, laboratory and other openings.
  • Regular length taper
  • Assorted sizes
The XXXX premium grade corks have fewer lenticels (crevices) than standard grade
Selected grade of fine textured corks. Sizes 22 through 30 have cemented construction.
This means they are better suited for use as laboratory corks and for transporting many kinds of liquids.