CellPro™ Neutralizing Solutions
Supplier: Alkali Scientific
DS0215 DB0319 DB0319-20L DS0215-4L
104026-840EA 1155.38 CAD
104026-840 104027-500 104027-480 104028-868
CellPro™ Neutralizing Solutions
Buffers Cell Culture Buffers
CellPro™ Neutralizing Solution is an essential reagent in molecular biology and biochemical experiments, particularly in protocols involving nucleic acid hybridization and blotting techniques.
  • Sterile
  • Filtered
This solution is specifically formulated to neutralize the effects of strong bases or acids used during the denaturation process. For example, after treating samples with denaturing solutions such as 0.5 M NaOH with 1.5 M NaCl, the CellPro™ Neutralizing Solution effectively restores the pH balance, creating an optimal environment for subsequent experimental steps.
Caution: UN No. 3248