Microcentrifuge Tube Boxes
Supplier: NEST SCIENTIFIC
613111 613001
77508-348CS 366.72 CAD
77508-348 77508-346
Microcentrifuge Tube Boxes
Boxes Boxes for Tubes
These boxes can be used for low temperature centralized storage of biological samples in microtubes.
  • Centralized storage of microtubes
  • Low temperature resistant
  • Deemed animal free
  • Non sterile
Clear polycarbonate cover for easy identification. Solid polypropylene base and polycarbonate cover. Withstand temperature ranging from –80 to 121 ℃.