Culture Tubes, 1.1 ml
Supplier: NEST SCIENTIFIC
628122 628124 628132 628134 628111 628112 628114
77508-470CS 498.77 CAD
77508-470 77508-472 77508-476 77508-478 77508-462 77508-464 77508-466
Culture Tubes, 1.1 ml
Tubes Culture Tubes
NEST 1.1 ml cluster tube box is compatible with multi-channel pipettes and automated liquid workstations.
The raw material of tube body is imported PP, which meets the USP-CLASS 6 standard. It has high transparency and good chemical stability, and is suitable for low-temperature storage of cells, drugs, tissues, sera, and other reagents.
NEST 1.1 ml cluster tubes are available in single tube, 8-strip, and 12-strip, and the tube caps are available in 8-strip and 12-strip for customers to choose from. They are compatible with multi-channel pipettes and automated liquid workstations with SBS standard size carriers.
The one-way cap design, with letter and number grids, ensures positive identification of samples. The bottom and side walls of the box holder are hollowed out for good thermal conductivity, and the box supports direct immersion in a water bath.
The raw material is imported PP, which conforms to USP class 6 standards. The 8-strip and 12-strip caps are made of TPE, which has better sealing performance. Resistant to low temperature storage at −80 °C and autoclavable.