High-Efficient Erlenmeyer Flasks With Tube
Supplier: NEST SCIENTIFIC
C10422-AZB090A C10423-BZC090A C10122-AZB050A C10123-BZC050A C10322-AZB050A C10322-AZB050B C10323-BZC050A C10442-AKA060A C20122-AZB050A C20222-AZB050A C20322-AZB050A C10222-AZB050A C10223-BZC050A
77509-070CS 1300.83 CAD
77509-070 77509-074 77509-000 77509-004 77509-046 77509-048 77509-050 77509-090 77509-092 77509-094 77509-096 77509-022 77509-026
High-Efficient Erlenmeyer Flasks With Tube
Flasks Erlenmeyer Flasks
The high-efficient Erlenmeyer flask (with tube) is designed to eliminate pipeline design, assembly, sterilization and other operations and improving production efficiency. It is ideal for shaker culture applications and can be used for scale-up culture of suspension cells.
  • Polycarbonate
  • HDPE cap
  • Bi-directional transfer cap with TPE tube
  • Handle: Nylon
  • Sterilized by E-beam, Sterility assurance level: SAL= 10⁻⁶
  • Non-pyrogenic and DNase/RNase free
  • Pipe diameter: ¹/₈" Int.Ø, ¹/₄" Ext.Ø
High-efficiency, large-volume culture flasks allow cells to show strongMore Product Information