Drum Tray, Eagle Manufacturing
Supplier: Eagle Manufacturing
1614 1618
56617-343EA 517.4 CAD
56617-343 56618-538
Drum Tray, Eagle Manufacturing
Drum Storage
Tray with reservoir keeps container away from spills
Ideal for use under drums during dispensing/filling operations. Holds up to 34L (9gal.) of spills when used with a 208.2L (55gal.) drum. Constructed of high-density polyethylene. Use drum tray dolly to easily transport drums between work areas without compromising spill protection.