PIG® Blue Absorbent Sock, New Pig
Supplier: New Pig
PIG201 4048 PIG202 PIG217 MA1097 2048
46000-530PK 289.66 CAD
46000-530 56620-002 56620-004 56620-010 56620-566 56620-588
PIG® Blue Absorbent Sock, New Pig
Spill Control Materials
PIG’s heaviest absorbent sock forms a barrier to prevent spills from spreading.
  • Heavy-duty socks hug floors to absorb spills and stop them from spreading
  • Tubular, dense construction has fine-grade vermiculite filler for superior diking
  • Absorb oils, coolants, solvents, and water
  • Stitch-bonded polypropylene skin is chemical- and burst-resistant
Extra-dense construction hugs floors and stays in place while containing spills. Fine-grade vermiculite filler quickly absorbs leaks, drips and spills; great for spill response or absorbing machine leaks. Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated.
Absorbs and retains oils, coolants, solvents and water. Excellent for diking and damming liquids. Can be air-dried for reuse when soaked with water.