Chromatography Needles, Point Style 2, Hamilton
Supplier: Hamilton
90022 90024 90039 90026 90030 90122 90138 90125 90139 90031 90033 90128 90131 90124 90126
60378-000PK 172.39 CAD
60378-000 60378-010 60378-021 60378-043 60378-087 60379-239 60379-250 60379-296 60379-352 60378-101 60378-123 60379-396 60379-432 89221-168 89221-170
Chromatography Needles, Point Style 2, Hamilton
Syringe Needles
For use on Luer and Luer-lock syringes.
This point style is recommended for most applications requiring the puncturing of a septum. Needle is cut at a 22° bevel with the tip of the needle slightly bent, partially obstructing the orifice, to prevent septum coring.
An “S” after needle gauge denotes a small bore. A needle with a small bore carries less dead volume than a standard bore, and has a thicker wall which can be an advantage in piercing septa.