Cubis® II Balances
Supplier: Sartorius
MCA125S-I-3-MTS MCA225S-I-3-MTS MCA226S-I-3-MTS MCE125S-I-3-MTS MCE225S-I-3-MTS MCA125S-U-MTS MCA8202S-0-MTS MCE8202S-0-MTS
CA77591-718EA 27132.42 CAD
CA77591-718 CA77591-720 CA77591-722 CA77591-724 CA77591-726 CA77547-558 CA77547-560 CA77547-562
Cubis® II Balances
Balances and Scales
The modular weighing system Cubis® II can be individually configured from different components. The combinability of the display unit, weighing module, draft shield, software packages for various applications and functions and a comprehensive range of accessories allows the individual adaptation of the Cubis® II balance to all weighing tasks.
  • Leading performance
  • Monolithical weighing system, integrated climate sensors, individual sample holders
  • Error-free operation
  • Individual QApp workflows
  • Motorized auto-leveling
  • Full end-to-end data integrity
  • Integrated audit-trail, state-of-the-art user management
  • Outstanding service support
  • Integrated status center, integrated service functions preventive maintenance on accredited standards
The Cubis® II range of premium laboratory balances with a maximum load between 2.1 g and 70 kg and a readability between 0.1 μg to 1 g provide the ideal model for every application.
Connection type: D-sub connector, 9-pin
Display type: Color touchscreen
Certifications: 21 CFR Part 11 compliance.