Glassware Carts, Labconco®
Supplier: Labconco
8032500 8045000
82006-868EA 1659 CAD
82006-868 82006-870
Glassware Carts, Labconco®
Trolleys
Designed for use with glassware and small equipment
Carts have a welded tubular steel frame with tough, corrosion-resistant glacier white epoxy coating, four 10.2cm (4") diameter casters (two with toe locks), two handles with ergonomic extruded rubber grips, and a heavy-duty removable plastic drip pan. Interchangeable wire baskets are vinyl-coated for glassware safety and easy storage.
Ordering information: Manufacturer's lifetime warranty.