Medium-Duty Open Tray Trucks, Lakeside®
Supplier: Lakeside
465 463 461 451 448 446 357 354 352
19758-324EA 15526.74 CAD
19758-324 19758-326 19758-328 19758-330 19758-332 19758-334 19758-336 19758-338 19758-340
Medium-Duty Open Tray Trucks, Lakeside®
Trolleys
These durable and versatile trucks are ideal for efficient transfer of any kind of maintenance, supply, or bulk item
Note: Capacity is the total cart capacity across all of the shelves with the weight evenly dispersed.
Trucks are equipped with two 12.7cm (5") rubber fixed casters and two 12.7cm (5") rubber swivel casters for quiet maneuverability and feature a recessed handle with wraparound strip bumper. Each shelf has one edge down and three up.
Ordering information: Trucks are shipped fully assembled. Custom shelf configurations are available; contact your Avantor sales representative for more information.