pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
BDH35309.606 BDH35310.601 BDH35311.604 BDH35312.607 92180.2047 92190.2047 BDH35316.601 BDH35317.604 92160.2047 BDH83930.601 92122.2047 BDH83932.601 BDH83933.601
BDH35309.606CS 18993.34 CAD
BDH35309.606 BDH35310.601 BDH35311.604 BDH35312.607 BDH35314.604 BDH35315.607 BDH35316.601 BDH35317.604 BDH35318.607 BDH83930.601 BDH83931.601 BDH83932.601 BDH83933.601
pH Test Strips, VWR Chemicals BDH®
Test Papers pH Indicator Papers and Test Strips
Used for rapid pH testing directly at the point of interest, strips are always ready for immediate use.
  • Designed for safe testing of dangerous, poisonous, or aggressive liquids
  • Long handle protects user from contact with sample
  • Fixed dye cannot bleed into the sample, protecting it against contamination and enabling measurements even in weakly buffered or strongly alkaline solutions
Dimensions: (LxW) 85x6 mm.
Ordering information: Strips are packaged 100 to a box. Included color chart has 4 different color blocks for each pH value