Whatman™ pH Indicator Papers, on Reels, Whatman products (Cytiva)
Supplier: Cytiva
2600-100A 2600-101A 2600-102A 2600-103A 2611-628 2611-628 2600-104A 2600-104A 2600-100A 2600-101A 2600-102A 2600-103A
14217-670PK 79.86 CAD
14217-670 14217-672 14217-674 14217-676 CA11008-596 11008-596 CA11013-070 10035-342 CA14217-670 CA14217-672 CA14217-674 CA14217-676
Whatman™ pH Indicator Papers, on Reels, Whatman products (Cytiva)
Test Papers pH Indicator Papers and Test Strips
Can be used for the rapid determination of pH values and has led to many applications in laboratories and industry
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Inexpensive
  • Convenient and portable for field use
Three colors roll: The paper has three separate indicator dye colour bands. The individual combination of colour change resulting from each test is compared with the colour-coded comparison chart printed on the dispenser, giving improved speed and accuracy in reading.
Available as standard or three colour ranges as rolls in a handy dispenser.