pH Indicator Paper DoubleZone, VWR Chemicals BDH®
Supplier: BDH
90301.2047 90302.2047 90303.2047 90304.2047 90305.2047 90306.2047
BDH35300.606CS 1616.11 CAD
BDH35300.606 BDH35301.600 BDH35302.603 BDH35303.606 BDH70035.607 BDH70036.601
pH Indicator Paper DoubleZone, VWR Chemicals BDH®
Test Papers pH Indicator Papers and Test Strips
These indicator papers show two different colors for each pH value at intervals of 0.3–1 pH unit, increasing the accuracy of readings and allowing good estimation of intermediate values
Paper comes on a plastic reel that will protect from many environmental influences, ensuring that the paper is always ready to use. Width: 10mm. Length: 5m.