Bransonic® Ultrasonic Pipet Cleaner, Branson
Supplier: Branson
Bransonic®
951-030
CA21812-750EA 5806.31 CAD
CA21812-750
Bransonic® Ultrasonic Pipet Cleaner, Branson
Ultrasonic Cleaners
For cleaning sampling pipets ultrasonically
Lipids and proteins are removed from the capillary bore of the pipet through the cavitation process.
Ordering information: Stainless steel cover and pipet basket sold separately.