AMCO Clear® Primary Turbidity Standards, GFS Chemicals
Supplier: GFS CHEMICALS INC
85151 85047 85040 85034 85075 85007 85011 85016 85024 85027 85031 85043 85050 85056 85062 85068 85074 85093 85123 85141 85159 86110 85009 85005 85014 85018 85030 85026 85051 85063 85069 85092 85122 85142 85150 85160 85164 85166
66115-038EA 218.94 CAD
66115-038 66115-136 66115-138 66115-140 66115-152 66115-000 66115-002 66115-004 66115-006 66115-008 66115-010 66115-014 66115-016 66115-018 66115-020 66115-022 66115-024 66115-026 66115-028 66115-034 66115-042 66115-046 66115-128 66115-130 66115-132 66115-134 66115-142 66115-144 66115-146 66115-148 66115-150 66115-154 66115-156 66115-162 66115-166 66115-170 66115-172 66115-174
AMCO Clear® Primary Turbidity Standards, GFS Chemicals
Standards Turbidity Standards
EPA-approved primary turbidity standards for calibrating turbidimeters and nephelometers
One year shelf life.
Ready to use at their stated value without mixing or diluting. Composed of submicron solid polymer beads suspended in ultra-pure water. Guaranteed accuracy within 1%. Safe, non-toxic, non-carcinogenic alternative to Formazin. Available for non-ratio and ratio meters.
Certifications: Approved by EPA, Standard Methods, ASTM, ASBC, and EBC. Traceable to NIST 1690, 1691.