UVP Blak-Ray® and Sterilizing UV Bench/Display Lamps, Analytik Jena
Supplier: Analytik Jena US
95-0045-01 95-0045-02 95-0045-04 95-0045-05 95-0045-07 95-0045-08 95-0043-01 95-0043-02 95-0043-04 95-0043-05 95-0043-07 95-0043-08 95-0042-06
21474-672EA 1104.91 CAD
21474-672 21474-674 21474-676 21474-678 21474-680 21474-682 21474-684 21474-686 21474-688 21474-690 21474-692 21474-694 36575-048 36575-059
UVP Blak-Ray® and Sterilizing UV Bench/Display Lamps, Analytik Jena
Lights UV Lights, Lamps and Bulbs
UV Bench/Display lamps combine streamlined design and simple, dependable operation. Blak-Ray® display lamps provide high intensity 365 nm radiation. The XX-15L model is designed for UV curing and photochemical applications where filtering of ambient light is not required. The BLB models use two 15 W, 20 W, or 40 W self-filtering BLB tubes for creating fluorescent displays or for non-destructive testing.
Blak-Ray® and sterilizing lamps feature on/off switch side-mounted on lamp
All 115 V models are ETL and ETLc approved. All 230 V models are CE approved.
Sterilizing bench lampsMore Product Information