PYREX® Polyethylene [ST] Stoppers, Corning
Supplier: Corning
7624-9 7624-16 7624-19 7624-32 7624-38 7624-8 7624-13 7624-22 7624-27
59492-041CS 50.24 CAD
59492-041 59492-085 59492-100 59492-125 59492-187 59492-039 59492-063 59492-121 59492-123
PYREX® Polyethylene [ST] Stoppers, Corning
Stoppers PE/LDPE/HDPE Stoppers
These PYREX® No.13 stoppers are made from linear HDPE and have closed bottoms to conform to Standard Taper stopper dimensions.
  • Blue and white with a closed bottom
  • Three rings on the plug provide an efficient seal
  • Improved design fully protects the neck from accidental tipping