Polyethylene [ST] Stoppers, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
28160R-9 28160R-13 28160R-16 28160R-19 28160R-22 28160R-27
89000-797CS 59.58 CAD
89000-797 89000-799 89000-801 89000-803 89000-805 89000-807
Polyethylene [ST] Stoppers, Kimble Chase
Stoppers PE/LDPE/HDPE Stoppers
For items ground to [ST] stopper dimensions
Stoppers are yellow and blue with a closed bottom. Made from linear high density polyethylene. Enlarged flange is designed to protect the neck if the glassware item is tipped over.