SmartTray™ System, Metro™
Supplier: Metropolitan Wire
CBNTC20MSOL1 CBNTC30MSOL1 CBNTC20MSOL2 CBNTC30MSOL2 CBNTC20M CBNTC30M CBNTCS20M
CA82002-428EA 7012.79 CAD
CA82002-428 CA82002-430 CA82002-434 CA82002-436 CA82002-446 CA82002-448 CA82002-450
SmartTray™ System, Metro™
Trolleys
Designed to maximize productivity by accelerating throughput and minimizing PCB handling damage
Premium ESD tray inlay (22221-396) offers maximum ESD protection in addition to superior resistance to heat, abrasion, and chemicals. The soft dissipative top layer makes it ideal for use in soldering and assembly areas. Economy tray inlays are not recommended for use in soldering applications.
This cart and tray system facilitates efficient tracking of work-in-process and finished goods with the use of bar code technology. Carts and trays can also be color-coded for easy sorting andMore Product Information